missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Methylphenoxy)piperidine, 97%, Thermo Scientific Chemicals

Product Code. 11499690
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-(3-Methylphenoxy)piperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(3-Methylphenoxy)piperidine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63843-46-9
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD06248345
InChI Key RTSXWJCYCFVWBU-UHFFFAOYSA-N
Synonym 4-3-methylphenoxy piperidine, 4-m-tolyloxy piperidine, acmc-1b64o
PubChem CID 17196730
IUPAC Name 4-(3-methylphenoxy)piperidine
SMILES CC1=CC(=CC=C1)OC2CCNCC2

Specifications

Melting Point 190°C to 191°C
Quantity 250 mg
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 4-(3-Methylphenoxy)piperidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'