Learn More
3-(Methoxycarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™
80.99 € - 259.00 €
Chemical Identifiers
CAS | 850567-26-9 |
---|---|
Molecular Formula | C8H10BNO4 |
Molecular Weight (g/mol) | 194.98 |
MDL Number | MFCD04115698 |
InChI Key | NDJZQOMZZXSXEM-UHFFFAOYSA-N |
Synonym | 3-methoxycarbamoyl phenyl boronic acid, 3-methoxycarbamoyl phenylboronic acid, acmc-209q0e, 3-borono-n-methoxybenzamide, boronic acid,b-3-methoxyamino carbonyl phenyl, 3-o-methylhydroxylaminocarbonyl benzeneboronic acid, 3-o-methylhydroxylaminocarbonyl phenyl boronic acid |
PubChem CID | 44118753 |
IUPAC Name | [3-(methoxycarbamoyl)phenyl]boronic acid |
SMILES | CONC(=O)C1=CC=CC(=C1)B(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsBenzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be a effective catalyst for amidation and esterification of carboxylic acids.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents. Keep container tightly closed.
Chemical Identifiers
850567-26-9 | |
194.98 | |
NDJZQOMZZXSXEM-UHFFFAOYSA-N | |
44118753 | |
CONC(=O)C1=CC=CC(=C1)B(O)O |
C8H10BNO4 | |
MFCD04115698 | |
3-methoxycarbamoyl phenyl boronic acid, 3-methoxycarbamoyl phenylboronic acid, acmc-209q0e, 3-borono-n-methoxybenzamide, boronic acid,b-3-methoxyamino carbonyl phenyl, 3-o-methylhydroxylaminocarbonyl benzeneboronic acid, 3-o-methylhydroxylaminocarbonyl phenyl boronic acid | |
[3-(methoxycarbamoyl)phenyl]boronic acid |
Specifications
850567-26-9 | |
MFCD04115698 | |
Slightly soluble in water. | |
CONC(=O)C1=CC=CC(=C1)B(O)O | |
194.98 | |
194.98 | |
3-(Methoxycarbamoyl)benzeneboronic acid |
C8H10BNO4 | |
3-methoxycarbamoyl phenyl boronic acid, 3-methoxycarbamoyl phenylboronic acid, acmc-209q0e, 3-borono-n-methoxybenzamide, boronic acid,b-3-methoxyamino carbonyl phenyl, 3-o-methylhydroxylaminocarbonyl benzeneboronic acid, 3-o-methylhydroxylaminocarbonyl phenyl boronic acid | |
NDJZQOMZZXSXEM-UHFFFAOYSA-N | |
[3-(methoxycarbamoyl)phenyl]boronic acid | |
44118753 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.