missing translation for 'onlineSavingsMsg'
Learn More

3-(Methoxycarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15439937
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Benzene boronic acid mediated the ortho specific hydroxyalkylation of phenols by aldehydes. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. It can be a effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-26-9
Molecular Formula C8H10BNO4
Molecular Weight (g/mol) 194.98
MDL Number MFCD04115698
InChI Key NDJZQOMZZXSXEM-UHFFFAOYSA-N
Synonym 3-methoxycarbamoyl phenyl boronic acid, 3-methoxycarbamoyl phenylboronic acid, acmc-209q0e, 3-borono-n-methoxybenzamide, boronic acid,b-3-methoxyamino carbonyl phenyl, 3-o-methylhydroxylaminocarbonyl benzeneboronic acid, 3-o-methylhydroxylaminocarbonyl phenyl boronic acid
PubChem CID 44118753
IUPAC Name [3-(methoxycarbamoyl)phenyl]boronic acid
SMILES CONC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 194.98
Percent Purity 97%
Chemical Name or Material 3-(Methoxycarbamoyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.