Learn More
3-Isopropoxybenzeneboronic acid, 97%, Thermo Scientific™
32.40 € - 493.00 €
Chemical Identifiers
CAS | 216485-86-8 |
---|---|
Molecular Formula | C9H13BO3 |
Molecular Weight (g/mol) | 180.01 |
MDL Number | MFCD03701512 |
InChI Key | UKZVUHVNTYDSOP-UHFFFAOYSA-N |
Synonym | 3-isopropoxyphenylboronic acid, 3-isopropoxyphenyl boronic acid, 3-isopropoxybenzeneboronic acid, 3-propan-2-yloxyphenyl boronic acid, 3-propan-2-yloxy phenyl boronic acid, 3-1-methylethoxy phenylboronic acid, 3-1-methylethyl oxyphenylboronic acid, 3-iso-propoxyphenylboronic acid, acmc-1ckjx, 5-isopropoxyphenylboronic acid |
PubChem CID | 5083932 |
IUPAC Name | (3-propan-2-yloxyphenyl)boronic acid |
SMILES | CC(C)OC1=CC=CC(=C1)B(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489747
|
Thermo Scientific Alfa Aesar
H52981.03 |
1 g |
32.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15499747
|
Thermo Scientific Alfa Aesar
H52981.06 |
5 g | N/A | ||||||
15409757
|
Thermo Scientific Alfa Aesar
H52981.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
216485-86-8 | |
180.01 | |
UKZVUHVNTYDSOP-UHFFFAOYSA-N | |
5083932 | |
CC(C)OC1=CC=CC(=C1)B(O)O |
C9H13BO3 | |
MFCD03701512 | |
3-isopropoxyphenylboronic acid, 3-isopropoxyphenyl boronic acid, 3-isopropoxybenzeneboronic acid, 3-propan-2-yloxyphenyl boronic acid, 3-propan-2-yloxy phenyl boronic acid, 3-1-methylethoxy phenylboronic acid, 3-1-methylethyl oxyphenylboronic acid, 3-iso-propoxyphenylboronic acid, acmc-1ckjx, 5-isopropoxyphenylboronic acid | |
(3-propan-2-yloxyphenyl)boronic acid |
Specifications
216485-86-8 | |
C9H13BO3 | |
3-isopropoxyphenylboronic acid, 3-isopropoxyphenyl boronic acid, 3-isopropoxybenzeneboronic acid, 3-propan-2-yloxyphenyl boronic acid, 3-propan-2-yloxy phenyl boronic acid, 3-1-methylethoxy phenylboronic acid, 3-1-methylethyl oxyphenylboronic acid, 3-iso-propoxyphenylboronic acid, acmc-1ckjx, 5-isopropoxyphenylboronic acid | |
CC(C)OC1=CC=CC(=C1)B(O)O | |
180.01 | |
180.01 | |
3-Isopropoxybenzeneboronic acid |
76°C to 81°C | |
MFCD03701512 | |
UKZVUHVNTYDSOP-UHFFFAOYSA-N | |
(3-propan-2-yloxyphenyl)boronic acid | |
5083932 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.