missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropoxybenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15499747
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216485-86-8
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD03701512
InChI Key UKZVUHVNTYDSOP-UHFFFAOYSA-N
Synonym 3-isopropoxyphenylboronic acid, 3-isopropoxyphenyl boronic acid, 3-isopropoxybenzeneboronic acid, 3-propan-2-yloxyphenyl boronic acid, 3-propan-2-yloxy phenyl boronic acid, 3-1-methylethoxy phenylboronic acid, 3-1-methylethyl oxyphenylboronic acid, 3-iso-propoxyphenylboronic acid, acmc-1ckjx, 5-isopropoxyphenylboronic acid
PubChem CID 5083932
IUPAC Name (3-propan-2-yloxyphenyl)boronic acid
SMILES CC(C)OC1=CC=CC(=C1)B(O)O

Specifications

Melting Point 76°C to 81°C
Quantity 5 g
Formula Weight 180.01
Percent Purity 97%
Chemical Name or Material 3-Isopropoxybenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.