missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyxanthone, 98%, Thermo Scientific Chemicals

69.30 € - 257.00 €

Chemical Identifiers

CAS 3722-51-8
Molecular Formula C13H8O3
Molecular Weight (g/mol) 212.204
MDL Number MFCD00604748
InChI Key XCJHDJAODLKGLG-UHFFFAOYSA-N
Synonym sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone
PubChem CID 5376013
IUPAC Name 3-hydroxyxanthen-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15460219
View Documents
Thermo Scientific Alfa Aesar
H63495.03
1 g
69.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
15470219
View Documents
Thermo Scientific Alfa Aesar
H63495.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3722-51-8
212.204
XCJHDJAODLKGLG-UHFFFAOYSA-N
5376013
C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O
C13H8O3
MFCD00604748
sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone
3-hydroxyxanthen-9-one

Specifications

3722-51-8
C13H8O3
170775
XCJHDJAODLKGLG-UHFFFAOYSA-N
3-hydroxyxanthen-9-one
5376013
98%
3-Hydroxyxanthone
243°C (decomposition)
MFCD00604748
sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone
C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O
212.204
212.2
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

missing translation for 'storageNote' : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only