missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxyxanthone, 98%, Thermo Scientific Chemicals

Product Code. 15470219
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3722-51-8
Molecular Formula C13H8O3
Molecular Weight (g/mol) 212.204
MDL Number MFCD00604748
InChI Key XCJHDJAODLKGLG-UHFFFAOYSA-N
Synonym sieber linker, 3-hydroxy-9h-xanthen-9-one, 3-hydroxy-xanthen-9-one, 3-hydroxyxanthone, 9h-xanthen-9-one, 3-hydroxy, sieber linker;, xanthen-9-one, 3-hydroxy, pubchem12819, acmc-209irq, 3-hydroxy-9-xanthenone
PubChem CID 5376013
IUPAC Name 3-hydroxyxanthen-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=C(O2)C=C(C=C3)O

Specifications

Melting Point 243°C (decomposition)
Quantity 5 g
Beilstein 170775
Formula Weight 212.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Hydroxyxanthone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'