Learn More
3-Fluoro-N-methylaniline, 97%, Thermo Scientific Chemicals
93.06 € - 1241.00 €
Chemical Identifiers
CAS | 1978-37-6 |
---|---|
Molecular Formula | C7H8FN |
Molecular Weight (g/mol) | 125.146 |
MDL Number | MFCD02683095 |
InChI Key | FHYDHJXZZQCXOX-UHFFFAOYSA-N |
Synonym | n-methyl-3-fluoroaniline, 3-fluoro-n-methylbenzenamine, 3-fluoro-n-methylanilin, n-methyl-m-fluoroaniline, acmc-209xf6, ksc497i4f, 3-fluoro-phenyl-methyl-amine, 3-fluoro-n-methylaniline, benzenamine, 3-fluoro-n-methyl |
PubChem CID | 2759011 |
IUPAC Name | 3-fluoro-N-methylaniline |
SMILES | CNC1=CC(=CC=C1)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11495137
|
Thermo Scientific Alfa Aesar
B22565.09 |
10 g |
93.06 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11405147
|
Thermo Scientific Alfa Aesar
B22565.18 |
50 g |
342.71 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415147
|
Thermo Scientific Alfa Aesar
B22565.30 |
250 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1978-37-6 | |
125.146 | |
FHYDHJXZZQCXOX-UHFFFAOYSA-N | |
2759011 | |
CNC1=CC(=CC=C1)F |
C7H8FN | |
MFCD02683095 | |
n-methyl-3-fluoroaniline, 3-fluoro-n-methylbenzenamine, 3-fluoro-n-methylanilin, n-methyl-m-fluoroaniline, acmc-209xf6, ksc497i4f, 3-fluoro-phenyl-methyl-amine, 3-fluoro-n-methylaniline, benzenamine, 3-fluoro-n-methyl | |
3-fluoro-N-methylaniline |
Specifications
1978-37-6 | |
94°C (19 mmHg) | |
Characteristic | |
MFCD02683095 | |
n-methyl-3-fluoroaniline, 3-fluoro-n-methylbenzenamine, 3-fluoro-n-methylanilin, n-methyl-m-fluoroaniline, acmc-209xf6, ksc497i4f, 3-fluoro-phenyl-methyl-amine, 3-fluoro-n-methylaniline, benzenamine, 3-fluoro-n-methyl | |
CNC1=CC(=CC=C1)F | |
125.146 | |
125.15 | |
3-Fluoro-N-methylaniline |
1.11 | |
86°C (187°F) | |
C7H8FN | |
UN2810 | |
FHYDHJXZZQCXOX-UHFFFAOYSA-N | |
3-fluoro-N-methylaniline | |
2759011 | |
97% |
Safety and Handling
GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P210-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P370+P378q-P501c
H227-H302+H332-H311-H315-H319-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.