missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-N-methylaniline, 97%, Thermo Scientific Chemicals

Product Code. 11405147
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1978-37-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.146
MDL Number MFCD02683095
InChI Key FHYDHJXZZQCXOX-UHFFFAOYSA-N
Synonym n-methyl-3-fluoroaniline, 3-fluoro-n-methylbenzenamine, 3-fluoro-n-methylanilin, n-methyl-m-fluoroaniline, acmc-209xf6, ksc497i4f, 3-fluoro-phenyl-methyl-amine, 3-fluoro-n-methylaniline, benzenamine, 3-fluoro-n-methyl
PubChem CID 2759011
IUPAC Name 3-fluoro-N-methylaniline
SMILES CNC1=CC(=CC=C1)F

Specifications

Density 1.11
Boiling Point 94°C (19 mmHg)
Flash Point 86°C (187°F)
Odor Characteristic
Quantity 50 g
UN Number UN2810
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-N-methylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'