Learn More
3-Chlorothiophene, 98%, may contain up to 2% DMF, Thermo Scientific Chemicals
59.90 € - 705.51 €
Chemical Identifiers
CAS | 17249-80-8 |
---|---|
Molecular Formula | C4H3ClS |
Molecular Weight (g/mol) | 118.578 |
MDL Number | MFCD00043887 |
InChI Key | QUBJDMPBDURTJT-UHFFFAOYSA-N |
Synonym | thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h |
PubChem CID | 87017 |
IUPAC Name | 3-chlorothiophene |
SMILES | C1=CSC=C1Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11455297
|
Thermo Scientific Alfa Aesar
B22669.06 |
5 g |
59.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465297
|
Thermo Scientific Alfa Aesar
B22669.14 |
25 g |
222.11 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475297
|
Thermo Scientific Alfa Aesar
B22669.22 |
100 g |
705.51 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17249-80-8 | |
118.578 | |
QUBJDMPBDURTJT-UHFFFAOYSA-N | |
87017 | |
C1=CSC=C1Cl |
C4H3ClS | |
MFCD00043887 | |
thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h | |
3-chlorothiophene |
Specifications
17249-80-8 | |
137°C to 139°C | |
Unpleasant | |
1.552 | |
UN1992 | |
thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h | |
C1=CSC=C1Cl | |
118.578 | |
118.59 | |
may contain up to 2% DMF |
1.276 | |
37°C (98°F) | |
C4H3ClS | |
MFCD00043887 | |
105255 | |
QUBJDMPBDURTJT-UHFFFAOYSA-N | |
3-chlorothiophene | |
87017 | |
98% | |
3-Chlorothiophene |
Safety and Handling
GHS H Statement
H331-H360-H318-H302-H312-H315-H335
Toxic if inhaled.
May damage fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H318-H335-H350-H360D-H373
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only