missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothiophene, 98%, may contain up to 2% DMF, Thermo Scientific Chemicals

Product Code. 11465297
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17249-80-8
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.578
MDL Number MFCD00043887
InChI Key QUBJDMPBDURTJT-UHFFFAOYSA-N
Synonym thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h
PubChem CID 87017
IUPAC Name 3-chlorothiophene
SMILES C1=CSC=C1Cl

Specifications

Density 1.276
Boiling Point 137°C to 139°C
Flash Point 37°C (98°F)
Odor Unpleasant
Refractive Index 1.552
Quantity 25 g
UN Number UN1992
Beilstein 105255
Formula Weight 118.59
Percent Purity 98%
Assay may contain up to 2% DMF
Chemical Name or Material 3-Chlorothiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'