missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)trichlorosilane, 97%, Thermo Scientific™

21.00 € - 213.00 €

Chemical Identifiers

CAS 2550-06-3
MDL Number MFCD00013694
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11365129
View Documents
Thermo Scientific Alfa Aesar
A17770.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11375129
View Documents
Thermo Scientific Alfa Aesar
A17770.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2550-06-3
MFCD00013694

Specifications

181°C to 183°C
C3H6Cl4Si
MFCD00013694
1737851
3-chloropropyltrichlorosilane, trichloro 3-chloropropyl silane, 3-chloropropyl trichlorosilane, chloropropyltrichlorosilane, silane, trichloro 3-chloropropyl, silane, 3-chloropropyltrichloro, trichloro-3-chloro-propyl-silane, silane, trichloro chloropropyl, g-chloropropyltrichlorosilane, gamma-chloropropyl trichlorosilane
C(C[Si](Cl)(Cl)Cl)CCl
211.97
211.98
Acrid
2550-06-3
Cl(CH2)3SiCl3
UN2987
84°C (183°F)
OOXSLJBUMMHDKW-UHFFFAOYSA-N
trichloro(3-chloropropyl)silane
75693
97%
(3-Chloropropyl)trichlorosilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.