missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)trichlorosilane, 97%, Thermo Scientific™

Product Code. 11365129
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-06-3
MDL Number MFCD00013694

Specifications

Boiling Point 181°C to 183°C
Quantity 25 g
Molecular Formula C3H6Cl4Si
Linear Formula Cl(CH2)3SiCl3
UN Number UN2987
Beilstein 1737851
Flash Point 84°C (183°F)
Synonym 3-chloropropyltrichlorosilane, trichloro 3-chloropropyl silane, 3-chloropropyl trichlorosilane, chloropropyltrichlorosilane, silane, trichloro 3-chloropropyl, silane, 3-chloropropyltrichloro, trichloro-3-chloro-propyl-silane, silane, trichloro chloropropyl, g-chloropropyltrichlorosilane, gamma-chloropropyl trichlorosilane
InChI Key OOXSLJBUMMHDKW-UHFFFAOYSA-N
SMILES C(C[Si](Cl)(Cl)Cl)CCl
IUPAC Name trichloro(3-chloropropyl)silane
Molecular Weight (g/mol) 211.97
PubChem CID 75693
Formula Weight 211.98
Percent Purity 97%
Odor Acrid
Chemical Name or Material (3-Chloropropyl)trichlorosilane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.