missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluoro-4-methoxyphenol, 97%, Thermo Scientific™

49.00 € - 176.00 €

Chemical Identifiers

CAS 1017777-55-7
Molecular Formula C7H6ClFO2
Molecular Weight (g/mol) 176.571
MDL Number MFCD09832264
InChI Key GHNZALBLIAFCPB-UHFFFAOYSA-N
PubChem CID 46737504
IUPAC Name 3-chloro-5-fluoro-4-methoxyphenol
SMILES COC1=C(C=C(C=C1Cl)O)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15434437
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H31801.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15424437
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H31801.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1017777-55-7
176.571
GHNZALBLIAFCPB-UHFFFAOYSA-N
3-chloro-5-fluoro-4-methoxyphenol
C7H6ClFO2
MFCD09832264
46737504
COC1=C(C=C(C=C1Cl)O)F

Specifications

1017777-55-7
C7H6ClFO2
GHNZALBLIAFCPB-UHFFFAOYSA-N
3-chloro-5-fluoro-4-methoxyphenol
46737504
97%
62°C to 65°C
MFCD09832264
COC1=C(C=C(C=C1Cl)O)F
176.571
176.57
3-Chloro-5-fluoro-4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications