missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluoro-4-methoxyphenol, 97%, Thermo Scientific™

Product Code. 15424437
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017777-55-7
Molecular Formula C7H6ClFO2
Molecular Weight (g/mol) 176.571
MDL Number MFCD09832264
InChI Key GHNZALBLIAFCPB-UHFFFAOYSA-N
PubChem CID 46737504
IUPAC Name 3-chloro-5-fluoro-4-methoxyphenol
SMILES COC1=C(C=C(C=C1Cl)O)F

Specifications

Melting Point 62°C to 65°C
Quantity 1 g
Formula Weight 176.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-fluoro-4-methoxyphenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'