Learn More
(±)-3-Butyn-2-ol, 98%, Thermo Scientific Chemicals
31.80 € - 465.00 €
Chemical Identifiers
CAS | 2028-63-9 |
---|---|
Molecular Formula | C4H6O |
Molecular Weight (g/mol) | 70.09 |
MDL Number | MFCD00004541 |
InChI Key | GKPOMITUDGXOSB-UHFFFAOYNA-N |
Synonym | 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol |
PubChem CID | 16239 |
IUPAC Name | but-3-yn-2-ol |
SMILES | CC(O)C#C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329509
|
Thermo Scientific Alfa Aesar
A11881.06 |
5 g |
31.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413854
|
Thermo Scientific Alfa Aesar
A11881.09 |
10 g |
52.30 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339509
|
Thermo Scientific Alfa Aesar
A11881.14 |
25 g |
95.80 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349509
|
Thermo Scientific Alfa Aesar
A11881.22 |
100 g |
264.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423854
|
Thermo Scientific Alfa Aesar
A11881.30 |
250 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2028-63-9 | |
70.09 | |
GKPOMITUDGXOSB-UHFFFAOYNA-N | |
16239 | |
CC(O)C#C |
C4H6O | |
MFCD00004541 | |
3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol | |
but-3-yn-2-ol |
Specifications
2028-63-9 | |
108°C to 109°C | |
C4H6O | |
MFCD00004541 | |
635722 | |
GKPOMITUDGXOSB-UHFFFAOYNA-N | |
but-3-yn-2-ol | |
16239 | |
98% |
0.894 | |
28°C (82°F) | |
1.426 | |
UN2929 | |
3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol | |
CC(O)C#C | |
70.09 | |
70.09 | |
(±)-3-Butyn-2-ol |
Safety and Handling
GHS H Statement
H300-H310-H226-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H300+H310+H330-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 217-978-1
missing translation for 'rtecsNumber' : ES0709800
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Self-reactive substance or mixture Type G
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only