missing translation for 'onlineSavingsMsg'
Learn More

(±)-3-Butyn-2-ol, 98%, Thermo Scientific Chemicals

Product Code. 11349509
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2028-63-9
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00004541
InChI Key GKPOMITUDGXOSB-UHFFFAOYNA-N
Synonym 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol
PubChem CID 16239
IUPAC Name but-3-yn-2-ol
SMILES CC(O)C#C

Specifications

Density 0.894
Boiling Point 108°C to 109°C
Flash Point 28°C (82°F)
Refractive Index 1.426
Quantity 100 g
UN Number UN2929
Beilstein 635722
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material (±)-3-Butyn-2-ol
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Self-reactive substance or mixture Type G
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'