Learn More
3-Bromopropylboronic acid pinacol ester, 98%, Thermo Scientific Chemicals
50.15 € - 760.00 €
Chemical Identifiers
CAS | 124215-44-7 |
---|---|
Molecular Formula | C9H18BBrO2 |
Molecular Weight (g/mol) | 248.96 |
MDL Number | MFCD10567053 |
InChI Key | CHQXFJUKMDJWHO-UHFFFAOYSA-N |
Synonym | 2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane |
PubChem CID | 14836669 |
IUPAC Name | 2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | CC1(C)OB(CCCBr)OC1(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15493967
|
Thermo Scientific Alfa Aesar
H30646.03 |
1 g |
50.15 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15403977
|
Thermo Scientific Alfa Aesar
H30646.06 |
5 g |
175.88 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15413977
|
Thermo Scientific Alfa Aesar
H30646.14 |
25 g | N/A | ||||||
Description
Used as primary and secondary intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as primary and secondary intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
124215-44-7 | |
248.96 | |
CHQXFJUKMDJWHO-UHFFFAOYSA-N | |
14836669 | |
CC1(C)OB(CCCBr)OC1(C)C |
C9H18BBrO2 | |
MFCD10567053 | |
2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane | |
2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
124215-44-7 | |
61°C to 64°C (0.35 mmHg) | |
MFCD10567053 | |
Not miscible or difficult to mix with water. | |
CC1(C)OB(CCCBr)OC1(C)C | |
248.96 | |
248.96 | |
3-Bromopropylboronic acid pinacol ester |
1.177 | |
C9H18BBrO2 | |
2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane | |
CHQXFJUKMDJWHO-UHFFFAOYSA-N | |
2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
14836669 | |
98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
- 3-Bromopropylboronic acid pinacol ester
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 3-Bromopropylboronicacid pinacol ester
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.