missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromopropylboronic acid pinacol ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Used as primary and secondary intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as primary and secondary intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Used as primary and secondary intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
Boiling Point | 61°C to 64°C (0.35 mmHg) |
Quantity | 25 g |
Formula Weight | 248.96 |
Percent Purity | 98% |
Chemical Name or Material | 3-Bromopropylboronic acid pinacol ester |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction