missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals

30.20 € - 520.00 €

Chemical Identifiers

CAS 7450-57-9
Molecular Formula C7H9N3O2
Molecular Weight (g/mol) 167.168
MDL Number MFCD00017068
InChI Key KNKNIBYXWQUDIH-UHFFFAOYSA-N
Synonym 3-amino-4-hydroxybenzhydrazide, 3-amino-4-hydroxy benzhydrazide, 4-???-2-aminophenol, #, 3-amino-4-hydroxybenzoic acid hydrazide, 3-amino-4-hydroxy benzoic acid hydrazide, 3-amino-4-hydroxybenzoyl hydrazine, benzoic acid,3-amino-4-hydroxy-, hydrazide, 3-amino-4-hydroxy-benzoic acid hydrazide
PubChem CID 585879
IUPAC Name 3-amino-4-hydroxybenzohydrazide
SMILES C1=CC(=C(C=C1C(=O)NN)N)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11464553
View Documents
Thermo Scientific Alfa Aesar
A11538.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11474553
View Documents
Thermo Scientific Alfa Aesar
A11538.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11484553
View Documents
Thermo Scientific Alfa Aesar
A11538.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7450-57-9
167.168
KNKNIBYXWQUDIH-UHFFFAOYSA-N
585879
C1=CC(=C(C=C1C(=O)NN)N)O
C7H9N3O2
MFCD00017068
3-amino-4-hydroxybenzhydrazide, 3-amino-4-hydroxy benzhydrazide, 4-???-2-aminophenol, #, 3-amino-4-hydroxybenzoic acid hydrazide, 3-amino-4-hydroxy benzoic acid hydrazide, 3-amino-4-hydroxybenzoyl hydrazine, benzoic acid,3-amino-4-hydroxy-, hydrazide, 3-amino-4-hydroxy-benzoic acid hydrazide
3-amino-4-hydroxybenzohydrazide

Specifications

7450-57-9
C7H9N3O2
6503217
KNKNIBYXWQUDIH-UHFFFAOYSA-N
3-amino-4-hydroxybenzohydrazide
585879
98%
250°C to 252°C
MFCD00017068
3-amino-4-hydroxybenzhydrazide, 3-amino-4-hydroxy benzhydrazide, 4-???-2-aminophenol, #, 3-amino-4-hydroxybenzoic acid hydrazide, 3-amino-4-hydroxy benzoic acid hydrazide, 3-amino-4-hydroxybenzoyl hydrazine, benzoic acid,3-amino-4-hydroxy-, hydrazide, 3-amino-4-hydroxy-benzoic acid hydrazide
C1=CC(=C(C=C1C(=O)NN)N)O
167.168
167.17
3-Amino-4-hydroxybenzhydrazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only