missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals

Product Code. 11474553
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7450-57-9
Molecular Formula C7H9N3O2
Molecular Weight (g/mol) 167.168
MDL Number MFCD00017068
InChI Key KNKNIBYXWQUDIH-UHFFFAOYSA-N
Synonym 3-amino-4-hydroxybenzhydrazide, 3-amino-4-hydroxy benzhydrazide, 4-???-2-aminophenol, #, 3-amino-4-hydroxybenzoic acid hydrazide, 3-amino-4-hydroxy benzoic acid hydrazide, 3-amino-4-hydroxybenzoyl hydrazine, benzoic acid,3-amino-4-hydroxy-, hydrazide, 3-amino-4-hydroxy-benzoic acid hydrazide
PubChem CID 585879
IUPAC Name 3-amino-4-hydroxybenzohydrazide
SMILES C1=CC(=C(C=C1C(=O)NN)N)O

Specifications

Melting Point 250°C to 252°C
Quantity 5 g
Beilstein 6503217
Formula Weight 167.17
Percent Purity 98%
Chemical Name or Material 3-Amino-4-hydroxybenzhydrazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'