missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)piperidine, 97%, Thermo Scientific Chemicals

84.60 € - 295.00 €

Chemical Identifiers

CAS 384346-27-4
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.689
MDL Number MFCD08061075
InChI Key OXELOOCWZRBKSV-UHFFFAOYSA-N
Synonym 3-4-chlorophenoxy piperidine, 3-4-chloro-phenoxy-piperidine, acmc-20ankc, piperidine,3-4-chlorophenoxy, 4-chloro-1-3-piperidyloxy benzene
PubChem CID 3865138
IUPAC Name 3-(4-chlorophenoxy)piperidine
SMILES C1CC(CNC1)OC2=CC=C(C=C2)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11439670
View Documents
Thermo Scientific Alfa Aesar
H50935.MD
250 mg
84.60 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11449670
View Documents
Thermo Scientific Alfa Aesar
H50935.03
1 g
295.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

384346-27-4
211.689
OXELOOCWZRBKSV-UHFFFAOYSA-N
3865138
C1CC(CNC1)OC2=CC=C(C=C2)Cl
C11H14ClNO
MFCD08061075
3-4-chlorophenoxy piperidine, 3-4-chloro-phenoxy-piperidine, acmc-20ankc, piperidine,3-4-chlorophenoxy, 4-chloro-1-3-piperidyloxy benzene
3-(4-chlorophenoxy)piperidine

Specifications

384346-27-4
MFCD08061075
Soluble in water.
C1CC(CNC1)OC2=CC=C(C=C2)Cl
211.689
211.69
3-(4-Chlorophenoxy)piperidine
C11H14ClNO
3-4-chlorophenoxy piperidine, 3-4-chloro-phenoxy-piperidine, acmc-20ankc, piperidine,3-4-chlorophenoxy, 4-chloro-1-3-piperidyloxy benzene
OXELOOCWZRBKSV-UHFFFAOYSA-N
3-(4-chlorophenoxy)piperidine
3865138
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.