missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)piperidine, 97%, Thermo Scientific Chemicals

Product Code. 11439670
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 384346-27-4
Molecular Formula C11H14ClNO
Molecular Weight (g/mol) 211.689
MDL Number MFCD08061075
InChI Key OXELOOCWZRBKSV-UHFFFAOYSA-N
Synonym 3-4-chlorophenoxy piperidine, 3-4-chloro-phenoxy-piperidine, acmc-20ankc, piperidine,3-4-chlorophenoxy, 4-chloro-1-3-piperidyloxy benzene
PubChem CID 3865138
IUPAC Name 3-(4-chlorophenoxy)piperidine
SMILES C1CC(CNC1)OC2=CC=C(C=C2)Cl

Specifications

Quantity 250 mg
Solubility Information Soluble in water.
Formula Weight 211.69
Percent Purity 97%
Chemical Name or Material 3-(4-Chlorophenoxy)piperidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.