missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-(Trifluoromethyl)piperazine, 97%, Thermo Scientific Chemicals

102.00 € - 289.00 €

Chemical Identifiers

CAS 131922-05-9
Molecular Formula C5H9F3N2
Molecular Weight (g/mol) 154.136
MDL Number MFCD07373382
InChI Key YNEJOOCZWXGXDU-UHFFFAOYSA-N
Synonym 2-trifluoromethyl piperazine, 2-trifluoromethyl-piperazine, +/--2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl, -2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl-9ci, acmc-20aolo, 2trifluoromethylpiperazine, m-trifluoromethylpiperazine
PubChem CID 18519725
IUPAC Name 2-(trifluoromethyl)piperazine
SMILES C1CNC(CN1)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15415257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32524.MD
250 mg
102.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15405257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H32524.03
1 g
289.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

131922-05-9
154.136
YNEJOOCZWXGXDU-UHFFFAOYSA-N
18519725
C1CNC(CN1)C(F)(F)F
C5H9F3N2
MFCD07373382
2-trifluoromethyl piperazine, 2-trifluoromethyl-piperazine, +/--2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl, -2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl-9ci, acmc-20aolo, 2trifluoromethylpiperazine, m-trifluoromethylpiperazine
2-(trifluoromethyl)piperazine

Specifications

131922-05-9
C5H9F3N2
UN3259
2-trifluoromethyl piperazine, 2-trifluoromethyl-piperazine, +/--2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl, -2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl-9ci, acmc-20aolo, 2trifluoromethylpiperazine, m-trifluoromethylpiperazine
C1CNC(CN1)C(F)(F)F
154.136
154.13
(+/-)-2-(Trifluoromethyl)piperazine
94°C to 97°C
MFCD07373382
Air Sensitive
YNEJOOCZWXGXDU-UHFFFAOYSA-N
2-(trifluoromethyl)piperazine
18519725
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only