missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-(Trifluoromethyl)piperazine, 97%, Thermo Scientific Chemicals

Product Code. 15405257
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131922-05-9
Molecular Formula C5H9F3N2
Molecular Weight (g/mol) 154.136
MDL Number MFCD07373382
InChI Key YNEJOOCZWXGXDU-UHFFFAOYSA-N
Synonym 2-trifluoromethyl piperazine, 2-trifluoromethyl-piperazine, +/--2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl, -2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl-9ci, acmc-20aolo, 2trifluoromethylpiperazine, m-trifluoromethylpiperazine
PubChem CID 18519725
IUPAC Name 2-(trifluoromethyl)piperazine
SMILES C1CNC(CN1)C(F)(F)F

Specifications

Melting Point 94°C to 97°C
Quantity 1 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 154.13
Percent Purity 97%
Chemical Name or Material (+/-)-2-(Trifluoromethyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'