Learn More
Ismatec™ 2-Stop Tubing, Tygon MHLL
Chemically resistant, long-life tubing with a smooth inner bore.
Brand:  Ismatec™ SC0716
Product Code. 10145072
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Chemically resistant to acetone, MEK, and other aggressive solvents.
  • Long-life tubing.
  • Plasticizer-free.
  • Smooth inner bore.
  • Low sorption maintains fluid integrity.
  • Minimal adhesion and diffusion.
Specifications
Specifications
FDA 21 CFR 177.2600;US Pharmacopoea Class VI
0.38 mm
381 mm
6/Pk
I.D. 0.38 mm
Orange-Green
Tygon MHLL
−70°C
Acids: Excellent, Alkaline solutions: Excellent, Solvents: Excellent, Pressure: Not rec., Vacuum: Good, Viscous media: Good, Sterile media: Good
74°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ismatec™ 2-Stop Tubing, Tygon MHLL > I.D. 0.38 mm, Orange-Green