Learn More
Ismatec™ MS-CA 2/660 Base-plate Cassette Pump
Brand:  Ismatec™ ISM846
Product Code. 10712372
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
CE Compatibility (EN 61326-1/EN 61010-1)
Fixed-Speed
IP 30
14.5 psi (Differential)
Yes
40°C
88.9 mm
134.62 mm
3 stop color coded
No Display
0.083 to 26 mL/min.
60 rpm
Click-n-go Cassettes, Operating Manual, Power Cord
5°C
124.46 mm
115/230 VAC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ismatec™ MS-CA 2/660 Base-plate Cassette Pump > Flow Rate: 0.083 to 26mL/min.