missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpiperazine, 96%, Thermo Scientific Chemicals

114.00 € - 484.00 €

Chemical Identifiers

CAS 5271-26-1
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD01871362
InChI Key RIMRLBGNCLMSNH-UHFFFAOYSA-N
Synonym 2-phenyl-piperazine, piperazine, 2-phenyl, 2-phenylpiperazin, 3-phenylpiperazine, 2-phenyl piperazine, pubchem8559, acmc-20a0jb, rs-2-phenylpiperazine, 2-phenylpiperazine, +,--2-phenyl-piperazine
PubChem CID 250673
IUPAC Name 2-phenylpiperazine
SMILES C1CNC(CN1)C2=CC=CC=C2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11485880
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26502.06
5 g
114.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11495880
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26502.14
25 g
484.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5271-26-1
162.236
RIMRLBGNCLMSNH-UHFFFAOYSA-N
250673
C1CNC(CN1)C2=CC=CC=C2
C10H14N2
MFCD01871362
2-phenyl-piperazine, piperazine, 2-phenyl, 2-phenylpiperazin, 3-phenylpiperazine, 2-phenyl piperazine, pubchem8559, acmc-20a0jb, rs-2-phenylpiperazine, 2-phenylpiperazine, +,--2-phenyl-piperazine
2-phenylpiperazine

Specifications

5271-26-1
C10H14N2
UN3259
2-phenyl-piperazine, piperazine, 2-phenyl, 2-phenylpiperazin, 3-phenylpiperazine, 2-phenyl piperazine, pubchem8559, acmc-20a0jb, rs-2-phenylpiperazine, 2-phenylpiperazine, +,--2-phenyl-piperazine
C1CNC(CN1)C2=CC=CC=C2
162.236
162.24
2-Phenylpiperazine
83°C to 87°C
MFCD01871362
Air Sensitive
RIMRLBGNCLMSNH-UHFFFAOYSA-N
2-phenylpiperazine
250673
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only