missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpiperazine, 96%, Thermo Scientific Chemicals

Product Code. 11485880
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5271-26-1
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
MDL Number MFCD01871362
InChI Key RIMRLBGNCLMSNH-UHFFFAOYSA-N
Synonym 2-phenyl-piperazine, piperazine, 2-phenyl, 2-phenylpiperazin, 3-phenylpiperazine, 2-phenyl piperazine, pubchem8559, acmc-20a0jb, rs-2-phenylpiperazine, 2-phenylpiperazine, +,--2-phenyl-piperazine
PubChem CID 250673
IUPAC Name 2-phenylpiperazine
SMILES C1CNC(CN1)C2=CC=CC=C2

Specifications

Melting Point 83°C to 87°C
Quantity 5 g
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 162.24
Percent Purity 96%
Chemical Name or Material 2-Phenylpiperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'