missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxybiphenyl-2-carboxaldehyde, 97+%, Thermo Scientific™

28.00 € - 364.82 €

Chemical Identifiers

CAS 93465-26-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD03424639
InChI Key ZYNNXRUYVVQLOW-UHFFFAOYSA-N
Synonym 2'-methoxy-biphenyl-2-carbaldehyde, 2-2-methoxyphenyl benzaldehyde, 2'-methoxybiphenyl-2-carbaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carbaldehyde, 2'-methoxybiphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carboxaldehyde, 2-methoxy-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde #
PubChem CID 602528
IUPAC Name 2-(2-methoxyphenyl)benzaldehyde
SMILES COC1=CC=CC=C1C2=CC=CC=C2C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15479077
View Documents
Thermo Scientific Alfa Aesar
H52384.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15459077
View Documents
Thermo Scientific Alfa Aesar
H52384.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15469077
View Documents
Thermo Scientific Alfa Aesar
H52384.06
5 g
364.82 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

93465-26-0
212.248
ZYNNXRUYVVQLOW-UHFFFAOYSA-N
602528
COC1=CC=CC=C1C2=CC=CC=C2C=O
C14H12O2
MFCD03424639
2'-methoxy-biphenyl-2-carbaldehyde, 2-2-methoxyphenyl benzaldehyde, 2'-methoxybiphenyl-2-carbaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carbaldehyde, 2'-methoxybiphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carboxaldehyde, 2-methoxy-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde #
2-(2-methoxyphenyl)benzaldehyde

Specifications

93465-26-0
MFCD03424639
2'-methoxy-biphenyl-2-carbaldehyde, 2-2-methoxyphenyl benzaldehyde, 2'-methoxybiphenyl-2-carbaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carbaldehyde, 2'-methoxybiphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carboxaldehyde, 2-methoxy-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde #
COC1=CC=CC=C1C2=CC=CC=C2C=O
212.248
212.24
2'-Methoxybiphenyl-2-carboxaldehyde
C14H12O2
Air Sensitive
ZYNNXRUYVVQLOW-UHFFFAOYSA-N
2-(2-methoxyphenyl)benzaldehyde
602528
≥97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications