missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxybiphenyl-2-carboxaldehyde, 97+%, Thermo Scientific™

Product Code. 15469077
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93465-26-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD03424639
InChI Key ZYNNXRUYVVQLOW-UHFFFAOYSA-N
Synonym 2'-methoxy-biphenyl-2-carbaldehyde, 2-2-methoxyphenyl benzaldehyde, 2'-methoxybiphenyl-2-carbaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carbaldehyde, 2'-methoxybiphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carboxaldehyde, 2-methoxy-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde #
PubChem CID 602528
IUPAC Name 2-(2-methoxyphenyl)benzaldehyde
SMILES COC1=CC=CC=C1C2=CC=CC=C2C=O

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 212.24
Percent Purity ≥97%
Chemical Name or Material 2'-Methoxybiphenyl-2-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'