Learn More
2-Ethylcyclohexanone, 99%, Thermo Scientific Chemicals
80.40 € - 310.00 €
Chemical Identifiers
CAS | 4423-94-3 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.199 |
MDL Number | MFCD00066306 |
InChI Key | WKYYYUWKFPFVEY-UHFFFAOYSA-N |
Synonym | 2-ethylcyclohexanone, cyclohexanone, 2-ethyl, 2-ethyl-cyclohexanone, acmc-1advf, 2-ethyl-cyclohexan-1-one |
PubChem CID | 20474 |
IUPAC Name | 2-ethylcyclohexan-1-one |
SMILES | CCC1CCCCC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11415616
|
Thermo Scientific Alfa Aesar
B20748.06 |
5 g |
80.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425616
|
Thermo Scientific Alfa Aesar
B20748.14 |
25 g |
310.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
4423-94-3 | |
126.199 | |
WKYYYUWKFPFVEY-UHFFFAOYSA-N | |
20474 | |
CCC1CCCCC1=O |
C8H14O | |
MFCD00066306 | |
2-ethylcyclohexanone, cyclohexanone, 2-ethyl, 2-ethyl-cyclohexanone, acmc-1advf, 2-ethyl-cyclohexan-1-one | |
2-ethylcyclohexan-1-one |
Specifications
4423-94-3 | |
MFCD00066306 | |
Soluble in alcohol. Insoluble in water. | |
CCC1CCCCC1=O | |
126.199 | |
126.2 | |
2-Ethylcyclohexanone |
C8H14O | |
2-ethylcyclohexanone, cyclohexanone, 2-ethyl, 2-ethyl-cyclohexanone, acmc-1advf, 2-ethyl-cyclohexan-1-one | |
WKYYYUWKFPFVEY-UHFFFAOYSA-N | |
2-ethylcyclohexan-1-one | |
20474 | |
99% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.