missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylcyclohexanone, 99%, Thermo Scientific Chemicals

Product Code. 11425616
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4423-94-3
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00066306
InChI Key WKYYYUWKFPFVEY-UHFFFAOYSA-N
Synonym 2-ethylcyclohexanone, cyclohexanone, 2-ethyl, 2-ethyl-cyclohexanone, acmc-1advf, 2-ethyl-cyclohexan-1-one
PubChem CID 20474
IUPAC Name 2-ethylcyclohexan-1-one
SMILES CCC1CCCCC1=O

Specifications

Quantity 25 g
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 2-Ethylcyclohexanone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.