Learn More
2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals
40.10 € - 372.86 €
Chemical Identifiers
CAS | 2566-44-1 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.13 |
MDL Number | MFCD00040762 |
InChI Key | LUNMJRJMSXZSLC-UHFFFAOYSA-N |
Synonym | cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687 |
PubChem CID | 137642 |
SMILES | OCCC1CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317536
|
Thermo Scientific Alfa Aesar
L11080.MD |
250 mg |
40.10 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397526
|
Thermo Scientific Alfa Aesar
L11080.03 |
1 g |
107.54 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307536
|
Thermo Scientific Alfa Aesar
L11080.06 |
5 g |
372.86 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
2566-44-1 | |
86.13 | |
LUNMJRJMSXZSLC-UHFFFAOYSA-N | |
137642 |
C5H10O | |
MFCD00040762 | |
cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687 | |
OCCC1CC1 |
Specifications
2566-44-1 | |
47°C (116°F) | |
1.4355 | |
UN1987 | |
cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687 | |
LUNMJRJMSXZSLC-UHFFFAOYSA-N | |
2-cyclopropylethanol | |
137642 | |
96% |
137°C to 138°C | |
C5H10O | |
MFCD00040762 | |
2036028 | |
Miscible with water. | |
OCCC1CC1 | |
86.13 | |
86.13 | |
2-Cyclopropylethanol |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only