missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals

Product Code. 11307536
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2566-44-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00040762
InChI Key LUNMJRJMSXZSLC-UHFFFAOYSA-N
Synonym cyclopropaneethanol, 2-cyclopropylethan-1-ol, 2-cyclopropyl-ethanol, 2-hydroxyethyl cyclopropane, cyclopropaneethanol∼ 2-hydroxyethyl cyclopropane, 2-cyclopropyl ethanol, 2-cyclopropane ethanol, acmc-1cgpb, cyclopropylmethyl carbinol, intermediates-zcf02687
PubChem CID 137642
SMILES OCCC1CC1

Specifications

Boiling Point 137°C to 138°C
Flash Point 47°C (116°F)
Refractive Index 1.4355
Quantity 5 g
UN Number UN1987
Beilstein 2036028
Solubility Information Miscible with water.
IUPAC Name 2-cyclopropylethan-1-ol
Formula Weight 86.13
Percent Purity 96%
Chemical Name or Material 2-Cyclopropylethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'