Learn More
2-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
58.40 € - 661.00 €
Chemical Identifiers
CAS | 69360-26-5 |
---|---|
Molecular Formula | C7H4ClNO2S |
Molecular Weight (g/mol) | 201.624 |
MDL Number | MFCD00052341 |
InChI Key | NQAYCMBZPAARNO-UHFFFAOYSA-N |
Synonym | 2-cyanobenzene-1-sulfonyl chloride, 2-cyanobenzenesulphonyl chloride, 2-cyanophenylsulfonyl chloride, 2-cyanobenzensulfonyl chloride, 2-cyanobenzenesulfonylchloride, 2-cyano-benzenesulfonyl chloride, pubchem5107, cyanobenzenesulfonyl chloride, 2-cyanobenzensulfonylchloride, acmc-1b3m7 |
PubChem CID | 522347 |
IUPAC Name | 2-cyanobenzenesulfonyl chloride |
SMILES | C1=CC=C(C(=C1)C#N)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438696
|
Thermo Scientific Alfa Aesar
A16042.03 |
1 g |
58.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448696
|
Thermo Scientific Alfa Aesar
A16042.06 |
5 g |
195.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458696
|
Thermo Scientific Alfa Aesar
A16042.14 |
25 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69360-26-5 | |
201.624 | |
NQAYCMBZPAARNO-UHFFFAOYSA-N | |
522347 | |
C1=CC=C(C(=C1)C#N)S(=O)(=O)Cl |
C7H4ClNO2S | |
MFCD00052341 | |
2-cyanobenzene-1-sulfonyl chloride, 2-cyanobenzenesulphonyl chloride, 2-cyanophenylsulfonyl chloride, 2-cyanobenzensulfonyl chloride, 2-cyanobenzenesulfonylchloride, 2-cyano-benzenesulfonyl chloride, pubchem5107, cyanobenzenesulfonyl chloride, 2-cyanobenzensulfonylchloride, acmc-1b3m7 | |
2-cyanobenzenesulfonyl chloride |
Specifications
69360-26-5 | |
C7H4ClNO2S | |
UN3261 | |
2-cyanobenzene-1-sulfonyl chloride, 2-cyanobenzenesulphonyl chloride, 2-cyanophenylsulfonyl chloride, 2-cyanobenzensulfonyl chloride, 2-cyanobenzenesulfonylchloride, 2-cyano-benzenesulfonyl chloride, pubchem5107, cyanobenzenesulfonyl chloride, 2-cyanobenzensulfonylchloride, acmc-1b3m7 | |
C1=CC=C(C(=C1)C#N)S(=O)(=O)Cl | |
201.624 | |
201.63 | |
2-Cyanobenzenesulfonyl chloride |
65°C to 69°C | |
MFCD00052341 | |
2721588 | |
NQAYCMBZPAARNO-UHFFFAOYSA-N | |
2-cyanobenzenesulfonyl chloride | |
522347 | |
97% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RUO – Research Use Only