missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11438696
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69360-26-5
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.624
MDL Number MFCD00052341
InChI Key NQAYCMBZPAARNO-UHFFFAOYSA-N
Synonym 2-cyanobenzene-1-sulfonyl chloride, 2-cyanobenzenesulphonyl chloride, 2-cyanophenylsulfonyl chloride, 2-cyanobenzensulfonyl chloride, 2-cyanobenzenesulfonylchloride, 2-cyano-benzenesulfonyl chloride, pubchem5107, cyanobenzenesulfonyl chloride, 2-cyanobenzensulfonylchloride, acmc-1b3m7
PubChem CID 522347
IUPAC Name 2-cyanobenzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)C#N)S(=O)(=O)Cl

Specifications

Melting Point 65°C to 69°C
Quantity 1 g
UN Number UN3261
Beilstein 2721588
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 2-Cyanobenzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'