Learn More
2-Chloro-6-fluorophenol, 97%, Thermo Scientific™
42.90 € - 151.00 €
Chemical Identifiers
CAS | 2040-90-6 |
---|---|
Molecular Formula | C6H4ClFO |
Molecular Weight (g/mol) | 146.545 |
MDL Number | MFCD01631574 |
InChI Key | QIAQIYQASAWZPP-UHFFFAOYSA-N |
Synonym | 2-chloro-6-fluoro-phenol, 2-chloro-6-fluoro phenol, phenol, 2-chloro-6-fluoro, pubchem2853, 6-chlor-2-fluorphenol, acmc-1cqou, 6-chloro-2-fluorophenol, 2-chlor-6-fluorphenol;, 2-chloro-6-fluorophenol;, 2-chlor-6-fluor-phenol; |
PubChem CID | 2773710 |
IUPAC Name | 2-chloro-6-fluorophenol |
SMILES | C1=CC(=C(C(=C1)Cl)O)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2040-90-6 | |
146.545 | |
QIAQIYQASAWZPP-UHFFFAOYSA-N | |
2773710 | |
C1=CC(=C(C(=C1)Cl)O)F |
C6H4ClFO | |
MFCD01631574 | |
2-chloro-6-fluoro-phenol, 2-chloro-6-fluoro phenol, phenol, 2-chloro-6-fluoro, pubchem2853, 6-chlor-2-fluorphenol, acmc-1cqou, 6-chloro-2-fluorophenol, 2-chlor-6-fluorphenol;, 2-chloro-6-fluorophenol;, 2-chlor-6-fluor-phenol; | |
2-chloro-6-fluorophenol |
Specifications
2040-90-6 | |
Phenol-like | |
MFCD01631574 | |
1936441 | |
QIAQIYQASAWZPP-UHFFFAOYSA-N | |
2-chloro-6-fluorophenol | |
2773710 | |
97% |
61°C to 65°C | |
C6H4ClFO | |
UN3261 | |
2-chloro-6-fluoro-phenol, 2-chloro-6-fluoro phenol, phenol, 2-chloro-6-fluoro, pubchem2853, 6-chlor-2-fluorphenol, acmc-1cqou, 6-chloro-2-fluorophenol, 2-chlor-6-fluorphenol;, 2-chloro-6-fluorophenol;, 2-chlor-6-fluor-phenol; | |
C1=CC(=C(C(=C1)Cl)O)F | |
146.545 | |
146.55 | |
2-Chloro-6-fluorophenol |
Safety and Handling
GHS H Statement
H340-H361-H314-H318-H302-H317
May cause genetic defects.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302-H314-H317-H335-H340-H361f
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 433-890-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.