missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorophenol, 97%, Thermo Scientific™

Product Code. 11450907
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2040-90-6
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
MDL Number MFCD01631574
InChI Key QIAQIYQASAWZPP-UHFFFAOYSA-N
Synonym 2-chloro-6-fluoro-phenol, 2-chloro-6-fluoro phenol, phenol, 2-chloro-6-fluoro, pubchem2853, 6-chlor-2-fluorphenol, acmc-1cqou, 6-chloro-2-fluorophenol, 2-chlor-6-fluorphenol;, 2-chloro-6-fluorophenol;, 2-chlor-6-fluor-phenol;
PubChem CID 2773710
IUPAC Name 2-chloro-6-fluorophenol
SMILES C1=CC(=C(C(=C1)Cl)O)F

Specifications

Melting Point 61°C to 65°C
Odor Phenol-like
Quantity 1 g
UN Number UN3261
Beilstein 1936441
Formula Weight 146.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.