missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(4-piperidinyloxy)benzonitrile, 98%, Thermo Scientific™

67.30 € - 236.00 €

Chemical Identifiers

CAS 796600-10-7
Molecular Formula C12H13ClN2O
Molecular Weight (g/mol) 236.70
MDL Number MFCD08060977
InChI Key AZTMLAAAPWXHMV-UHFFFAOYSA-N
Synonym 2-chloro-4-4-piperidinyloxy benzonitrile, 2-chloro-4-piperidin-4-yloxy benzonitrile, acmc-20anzz, 2-chloro-4-piperidin-4-yl-oxy benzonitrile, benzonitrile,2-chloro-4-4-piperidinyloxy, 2-chloro-4-4-piperidyloxy benzenecarbonitrile, 2-chloro-4-piperidin-4-yl oxy benzonitrile, benzonitrile, 2-chloro-4-4-piperidinyloxy
PubChem CID 17986291
IUPAC Name 2-chloro-4-piperidin-4-yloxybenzonitrile
SMILES ClC1=C(C=CC(OC2CCNCC2)=C1)C#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494730
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50774.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11404740
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50774.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

796600-10-7
236.70
AZTMLAAAPWXHMV-UHFFFAOYSA-N
17986291
ClC1=C(C=CC(OC2CCNCC2)=C1)C#N
C12H13ClN2O
MFCD08060977
2-chloro-4-4-piperidinyloxy benzonitrile, 2-chloro-4-piperidin-4-yloxy benzonitrile, acmc-20anzz, 2-chloro-4-piperidin-4-yl-oxy benzonitrile, benzonitrile,2-chloro-4-4-piperidinyloxy, 2-chloro-4-4-piperidyloxy benzenecarbonitrile, 2-chloro-4-piperidin-4-yl oxy benzonitrile, benzonitrile, 2-chloro-4-4-piperidinyloxy
2-chloro-4-piperidin-4-yloxybenzonitrile

Specifications

796600-10-7
MFCD08060977
2-chloro-4-4-piperidinyloxy benzonitrile, 2-chloro-4-piperidin-4-yloxy benzonitrile, acmc-20anzz, 2-chloro-4-piperidin-4-yl-oxy benzonitrile, benzonitrile,2-chloro-4-4-piperidinyloxy, 2-chloro-4-4-piperidyloxy benzenecarbonitrile, 2-chloro-4-piperidin-4-yl oxy benzonitrile, benzonitrile, 2-chloro-4-4-piperidinyloxy
ClC1=C(C=CC(OC2CCNCC2)=C1)C#N
236.70
236.7
2-Chloro-4-(4-piperidinyloxy)benzonitrile
C12H13ClN2O
UN3439
AZTMLAAAPWXHMV-UHFFFAOYSA-N
2-chloro-4-piperidin-4-yloxybenzonitrile
17986291
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications