missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(4-piperidinyloxy)benzonitrile, 98%, Thermo Scientific™

Product Code. 11404740
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 796600-10-7
Molecular Formula C12H13ClN2O
Molecular Weight (g/mol) 236.70
MDL Number MFCD08060977
InChI Key AZTMLAAAPWXHMV-UHFFFAOYSA-N
Synonym 2-chloro-4-4-piperidinyloxy benzonitrile, 2-chloro-4-piperidin-4-yloxy benzonitrile, acmc-20anzz, 2-chloro-4-piperidin-4-yl-oxy benzonitrile, benzonitrile,2-chloro-4-4-piperidinyloxy, 2-chloro-4-4-piperidyloxy benzenecarbonitrile, 2-chloro-4-piperidin-4-yl oxy benzonitrile, benzonitrile, 2-chloro-4-4-piperidinyloxy
PubChem CID 17986291
IUPAC Name 2-chloro-4-piperidin-4-yloxybenzonitrile
SMILES ClC1=C(C=CC(OC2CCNCC2)=C1)C#N

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 236.7
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-(4-piperidinyloxy)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'