missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylthiophene, 98%, Thermo Scientific Chemicals

27.00 € - 158.00 €

Chemical Identifiers

CAS 135-00-2
Molecular Formula C11H8OS
Molecular Weight (g/mol) 188.244
MDL Number MFCD00022495
InChI Key DWYFUJJWTRPARQ-UHFFFAOYSA-N
Synonym 2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene
PubChem CID 67262
IUPAC Name phenyl(thiophen-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CS2
Mehr Spezifikationen ansehen

Produkte 3
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11319679
Unterlagen ansehen Aktionsdetails
Thermo Scientific Alfa Aesar
A12036.06
5 g
27.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11499902
Unterlagen ansehen Aktionsdetails
Thermo Scientific Alfa Aesar
A12036.09
10 g
42.80 €
10g
Please to purchase this item. Need a web account? Register with us today!
11409912
Unterlagen ansehen Aktionsdetails
Thermo Scientific Alfa Aesar
A12036.18
50 g
158.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

135-00-2
188.244
DWYFUJJWTRPARQ-UHFFFAOYSA-N
67262
C1=CC=C(C=C1)C(=O)C2=CC=CS2
C11H8OS
MFCD00022495
2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene
phenyl(thiophen-2-yl)methanone

Spezifikation

135-00-2
124°C to 126°C (0.5 mmHg)
MFCD00022495
2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene
C1=CC=C(C=C1)C(=O)C2=CC=CS2
188.244
188.25
2-Benzoylthiophene
55°C to 57°C
C11H8OS
3929
DWYFUJJWTRPARQ-UHFFFAOYSA-N
phenyl(thiophen-2-yl)methanone
67262
98%
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'einecsNumber' : 205-169-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

RUO – Research Use Only