missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoylthiophene, 98%, Thermo Scientific Chemicals

Product Code. 11409912
Click to view available options
Quantity:
5 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-00-2
Molecular Formula C11H8OS
Molecular Weight (g/mol) 188.244
MDL Number MFCD00022495
InChI Key DWYFUJJWTRPARQ-UHFFFAOYSA-N
Synonym 2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene
PubChem CID 67262
IUPAC Name phenyl(thiophen-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CS2

Specifications

Melting Point 55°C to 57°C
Boiling Point 124°C to 126°C (0.5 mmHg)
Quantity 50 g
Beilstein 3929
Formula Weight 188.25
Percent Purity 98%
Chemical Name or Material 2-Benzoylthiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'