Learn More
2-Acetyl-6-bromopyridine, 97%, Thermo Scientific Chemicals
20.00 € - 203.00 €
Chemical Identifiers
CAS | 49669-13-8 |
---|---|
Molecular Formula | C7H6BrNO |
Molecular Weight (g/mol) | 200.035 |
MDL Number | MFCD00272200 |
InChI Key | RUJTWTUYVOEEFW-UHFFFAOYSA-N |
Synonym | 2-acetyl-6-bromopyridine, 2-bromo-6-acetylpyridine, 1-6-bromopyridin-2-yl ethanone, 1-6-bromopyridin-2-yl ethan-1-one, 1-6-bromo-2-pyridinyl-1-ethanone, ethanone, 1-6-bromo-2-pyridinyl, bromopyridinylethanone, pubchem13351, acmc-1ap21, ksc490s1f |
PubChem CID | 11298578 |
IUPAC Name | 1-(6-bromopyridin-2-yl)ethanone |
SMILES | CC(=O)C1=NC(=CC=C1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442388
|
Thermo Scientific Alfa Aesar
H55981.MD |
250 mg | N/A | ||||||
15422388
|
Thermo Scientific Alfa Aesar
H55981.03 |
1 g |
56.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15432388
|
Thermo Scientific Alfa Aesar
H55981.06 |
5 g |
203.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.
Solubility
Soluble in methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
49669-13-8 | |
200.035 | |
RUJTWTUYVOEEFW-UHFFFAOYSA-N | |
11298578 | |
CC(=O)C1=NC(=CC=C1)Br |
C7H6BrNO | |
MFCD00272200 | |
2-acetyl-6-bromopyridine, 2-bromo-6-acetylpyridine, 1-6-bromopyridin-2-yl ethanone, 1-6-bromopyridin-2-yl ethan-1-one, 1-6-bromo-2-pyridinyl-1-ethanone, ethanone, 1-6-bromo-2-pyridinyl, bromopyridinylethanone, pubchem13351, acmc-1ap21, ksc490s1f | |
1-(6-bromopyridin-2-yl)ethanone |
Specifications
49669-13-8 | |
C7H6BrNO | |
2-acetyl-6-bromopyridine, 2-bromo-6-acetylpyridine, 1-6-bromopyridin-2-yl ethanone, 1-6-bromopyridin-2-yl ethan-1-one, 1-6-bromo-2-pyridinyl-1-ethanone, ethanone, 1-6-bromo-2-pyridinyl, bromopyridinylethanone, pubchem13351, acmc-1ap21, ksc490s1f | |
RUJTWTUYVOEEFW-UHFFFAOYSA-N | |
1-(6-bromopyridin-2-yl)ethanone | |
11298578 | |
97% |
51°C to 55°C | |
MFCD00272200 | |
Soluble in methanol. | |
CC(=O)C1=NC(=CC=C1)Br | |
200.035 | |
200.03 | |
2-Acetyl-6-bromopyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.