missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-6-bromopyridine, 97%, Thermo Scientific Chemicals

Product Code. 15442388
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetyl-6-bromopyridine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as OLED intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49669-13-8
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00272200
InChI Key RUJTWTUYVOEEFW-UHFFFAOYSA-N
Synonym 2-acetyl-6-bromopyridine, 2-bromo-6-acetylpyridine, 1-6-bromopyridin-2-yl ethanone, 1-6-bromopyridin-2-yl ethan-1-one, 1-6-bromo-2-pyridinyl-1-ethanone, ethanone, 1-6-bromo-2-pyridinyl, bromopyridinylethanone, pubchem13351, acmc-1ap21, ksc490s1f
PubChem CID 11298578
IUPAC Name 1-(6-bromopyridin-2-yl)ethanone
SMILES CC(=O)C1=NC(=CC=C1)Br

Specifications

Melting Point 51°C to 55°C
Quantity 250 mg
Solubility Information Soluble in methanol.
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 2-Acetyl-6-bromopyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.