Learn More
2,6-Dimethyl-γ-pyrone, 99%, Thermo Scientific Chemicals
126.00 € - 1774.00 €
Chemical Identifiers
CAS | 1004-36-0 |
---|---|
Molecular Formula | C7H8O2 |
Molecular Weight (g/mol) | 124.14 |
MDL Number | MFCD00006579 |
InChI Key | VSYFZULSKMFUJJ-UHFFFAOYSA-N |
Synonym | 2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601 |
PubChem CID | 13862 |
IUPAC Name | 2,6-dimethylpyran-4-one |
SMILES | CC1=CC(=O)C=C(O1)C |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
10725951
|
Thermo Scientific Acros
116740250 |
25 g | Glass bottle |
126.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10102290
|
Thermo Scientific Acros
116741000 |
100 g | Glass bottle |
429.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686544
|
Thermo Scientific Acros
116745000 |
500 g | Glass bottle | ||||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
1004-36-0 | |
124.14 | |
VSYFZULSKMFUJJ-UHFFFAOYSA-N | |
13862 | |
CC1=CC(=O)C=C(O1)C |
C7H8O2 | |
MFCD00006579 | |
2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601 | |
2,6-dimethylpyran-4-one |
Spezifikation
1004-36-0 | |
100.0 | |
White to Beige | |
Authentic | |
C7H8O2 | |
17, 291 | |
VSYFZULSKMFUJJ-UHFFFAOYSA-N | |
2,6-dimethylpyran-4-one | |
13862 | |
99% | |
2,6-Dimethyl-γ-pyrone |
98.5 | |
133°C to 137°C | |
248°C to 250°C | |
98.5% min. (GC) | |
MFCD00006579 | |
2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601 | |
CC1=CC(=O)C=C(O1)C | |
124.14 | |
124.14 | |
Crystalline Powder or Crystals |
Sicherheit und Handhabung
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 213-719-1
- 2,6-Dimethyl-y-pyrone
Signalwort
- Warning
Gefahrenkategorie
- Acute toxicity Category 4
Gefahrenhinweis
- H302-Harmful if swallowed.
Sicherheitshinweis
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only