missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethyl-γ-pyrone, 99%, Thermo Scientific Chemicals

Product Code. 10686544
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1004-36-0
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00006579
InChI Key VSYFZULSKMFUJJ-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601
PubChem CID 13862
IUPAC Name 2,6-dimethylpyran-4-one
SMILES CC1=CC(=O)C=C(O1)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 133°C to 137°C
Color White to Beige
Boiling Point 248°C to 250°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 500 g
Beilstein 17, 291
Formula Weight 124.14
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,6-Dimethyl-γ-pyrone
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dimethyl-y-pyrone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'