Learn More
2,5-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals
29.95 € - 274.37 €
Chemical Identifiers
CAS | 93-02-7 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
InChI Key | AFUKNJHPZAVHGQ-UHFFFAOYSA-N |
Synonym | benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde |
PubChem CID | 66726 |
IUPAC Name | 2,5-dimethoxybenzaldehyde |
SMILES | COC1=CC(=C(C=C1)OC)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10715761
|
Thermo Scientific Acros
115390250 |
25 g | Glass bottle |
29.95 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10499210
|
Thermo Scientific Acros
115391000 |
100 g | Glass bottle |
78.99 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924791
|
Thermo Scientific Acros
115395000 |
500 g | Glass bottle |
274.37 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-02-7 | |
166.18 | |
benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde | |
2,5-dimethoxybenzaldehyde |
C9H10O3 | |
AFUKNJHPZAVHGQ-UHFFFAOYSA-N | |
66726 | |
COC1=CC(=C(C=C1)OC)C=O |
Specifications
93-02-7 | |
100.0 | |
Yellow to Beige | |
113°C | |
96% min. (GC) | |
(CH3O)2C6H3CHO | |
benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde | |
AFUKNJHPZAVHGQ-UHFFFAOYSA-N | |
2,5-dimethoxybenzaldehyde | |
66726 | |
97% | |
2, 5-Dimethoxybenzaldehyde |
96.0 | |
45.0°C to 50.0°C | |
146.0°C (10.0 mmHg) | |
Authentic | |
C9H10O3 | |
08,245 | |
Solubility in water: 795mg/l (25°C). Other solubilities: soluble in most common organic solvents | |
COC1=CC(=C(C=C1)OC)C=O | |
166.18 | |
166.18 | |
Crystalline Powder, Crystals or Chunks |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-211-5
RUO – Research Use Only