missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 10499210
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-02-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
InChI Key AFUKNJHPZAVHGQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2,5-dimethoxy, 2,5-dimethoxy benzaldehyde, unii-w49s1ppl78, 2,5-dimethoxybenzald, pubchem2176, gentisic aldehyde, 5-dimethoxy benzaldehyde, 2,5-dimethoxybenzaldehyd, 2,5-dimethoxybenzaldehye, 2.5-dimethoxybenzaldehyde
PubChem CID 66726
IUPAC Name 2,5-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)OC)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 45.0°C to 50.0°C
Color Yellow to Beige
Boiling Point 146.0°C (10.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CHO
Quantity 100 g
Beilstein 08,245
Solubility Information Solubility in water: 795mg/l (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 166.18
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2, 5-Dimethoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'