missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde, 98%, Thermo Scientific Chemicals

87.70 € - 247.00 €

Chemical Identifiers

CAS 325461-35-6
Molecular Formula C28H46O4
Molecular Weight (g/mol) 446.672
MDL Number MFCD03427237
InChI Key JMACLBZCZVVLDQ-UHFFFAOYSA-N
Synonym 2,5-bis 3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctoxy terephthalaldehyde, 2,5-bis-3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctyl oxy benzene-1,4-dicarbaldehyde
PubChem CID 3653721
IUPAC Name 2,5-bis(3,7-dimethyloctoxy)terephthalaldehyde
SMILES CC(C)CCCC(C)CCOC1=CC(=C(C=C1C=O)OCCC(C)CCCC(C)C)C=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15429135
View Documents
Thermo Scientific Alfa Aesar
H51067.MD
250 mg
87.70 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15419135
View Documents
Thermo Scientific Alfa Aesar
H51067.03
1 g
247.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

325461-35-6
446.672
JMACLBZCZVVLDQ-UHFFFAOYSA-N
3653721
CC(C)CCCC(C)CCOC1=CC(=C(C=C1C=O)OCCC(C)CCCC(C)C)C=O
C28H46O4
MFCD03427237
2,5-bis 3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctoxy terephthalaldehyde, 2,5-bis-3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctyl oxy benzene-1,4-dicarbaldehyde
2,5-bis(3,7-dimethyloctoxy)terephthalaldehyde

Specifications

325461-35-6
MFCD03427237
JMACLBZCZVVLDQ-UHFFFAOYSA-N
2,5-bis(3,7-dimethyloctoxy)terephthalaldehyde
3653721
98%
C28H46O4
2,5-bis 3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctoxy terephthalaldehyde, 2,5-bis-3,7-dimethyloctyloxy terephthalaldehyde, 2,5-bis 3,7-dimethyloctyl oxy benzene-1,4-dicarbaldehyde
CC(C)CCCC(C)CCOC1=CC(=C(C=C1C=O)OCCC(C)CCCC(C)C)C=O
446.672
446.66
2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only